ENAH03A0225D Enamine

5-(1-{[(9H-fluoren-9-yl)methoxy]carbonyl}piperidin-4-yl)-1-methyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD31559173
  • Cas Number : 2137972-03-1
  • Molecular Weight : 432.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-658128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-658128
Availability details for
Quantity
  • Availability
  • Add to Cart