ENAH03A07CAF Enamine

4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine-3-carboxamide

  • MDL number MFCD20683729
  • Cas Number : 115931-34-5
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1119311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1119311
Availability details for
Quantity
  • Availability
  • Add to Cart