ENAH03A07DD6 Enamine

2-cyclopropyl-1H-indol-5-amine

  • MDL number MFCD20702033
  • Cas Number : 952664-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1119609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1119609
Availability details for
Quantity
  • Availability
  • Add to Cart