ENAH03A11A4C Enamine

1-methyl-3-(pyridin-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD08059967
  • Cas Number : 92406-50-3
  • Molecular Weight : 174.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1148087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1148087
Availability details for
Quantity
  • Availability
  • Add to Cart