ENAH03A135AD Enamine

4,4-dimethylpiperidine-1-carbonyl chloride

  • MDL number MFCD23790266
  • Cas Number : 1516586-68-7
  • Molecular Weight : 175.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1155041

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1155041
pictograms GHS05,GHS07
hstatements H302,H314,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart