ENAH03A1690C Enamine

6-bromopyrido[2,3-d]pyrimidin-2-amine

  • MDL number MFCD12400822
  • Cas Number : 882670-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1162559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1162559
Availability details for
Quantity
  • Availability
  • Add to Cart