ENAH03A18757 Enamine

2-(bromomethyl)-6-chlorobenzonitrile

  • MDL number MFCD18392460
  • Cas Number : 863676-23-7
  • Molecular Weight : 230.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1168841

Safety Information

signalword Danger
pcodes P260,P261,P264,P280,P285,P301+P330+P331,P303+P361+P353,P304+P340,P304+P341,P305+P351+P338,P310,P342+P311,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1168841
pictograms GHS05,GHS08
hstatements H314,H334
Availability details for
Quantity
  • Availability
  • Add to Cart