ENAH03A1A29E Enamine

5-amino-1,4-dimethyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD26669621
  • Cas Number : 1368364-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-729495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-729495
Availability details for
Quantity
  • Availability
  • Add to Cart