ENAH03A1A2B7 Enamine

2-cyclopropylimidazo[1,2-a]pyridine-6-carboxylic acid

  • MDL number MFCD19606477
  • Cas Number : 1343134-57-5
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1197804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1197804
Availability details for
Quantity
  • Availability
  • Add to Cart