ENAH03A1C345 Enamine

{4-methyl-2-oxabicyclo[2.1.1]hexan-1-yl}methanamine

  • MDL number MFCD31559483
  • Cas Number : 2138031-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-832810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-832810
Availability details for
Quantity
  • Availability
  • Add to Cart