ENAH03A1C473 Enamine

tert-butyl N-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]carbamate

  • MDL number MFCD26518862
  • Cas Number : 1823257-56-2
  • Molecular Weight : 269.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-818745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-818745
Availability details for
Quantity
  • Availability
  • Add to Cart