ENAH03A1C97F Enamine

(2E)-3-(4-butoxy-3-methoxyphenyl)prop-2-enoic acid

  • MDL number MFCD02256113
  • Cas Number : 115375-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-832974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-832974
Availability details for
Quantity
  • Availability
  • Add to Cart