ENAH03A1C9F4 Enamine

(2E)-3-{4-[1-(cyclopropylformamido)ethyl]-3-(trifluoromethyl)phenyl}prop-2-enoic acid

  • MDL number MFCD28348352

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-833058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-833058
Availability details for
Quantity
  • Availability
  • Add to Cart