ENAH03A1CA37 Enamine

(2E)-3-(1,5-dimethyl-1H-pyrazol-4-yl)prop-2-enoic acid

  • MDL number MFCD02090861
  • Cas Number : 689251-96-5
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-833084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-833084
Availability details for
Quantity
  • Availability
  • Add to Cart