ENAH03A1CF30 Enamine

tert-butyl 3a-(aminomethyl)-octahydro-1H-indole-1-carboxylate

  • MDL number MFCD31689394
  • Cas Number : 2138231-04-4
  • Molecular Weight : 254.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-841165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-841165
Availability details for
Quantity
  • Availability
  • Add to Cart