ENAH03A1DCFE Enamine

ethyl 3-amino-3-(4,4-difluorocyclohexyl)propanoate

  • MDL number MFCD31794423
  • Cas Number : 2138364-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-843552

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-843552
Availability details for
Quantity
  • Availability
  • Add to Cart