ENAH03A1DEDB Enamine

(2-cyclobutylphenyl)methanol

  • MDL number MFCD28516888
  • Cas Number : 1593904-09-6
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-843219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-843219
Availability details for
Quantity
  • Availability
  • Add to Cart