ENAH03BB2FCE Enamine

4,6-dichloro-N-phenyl-1,3,5-triazin-2-amine

  • MDL number MFCD00452914
  • Cas Number : 2272-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126334
Availability details for
Quantity
  • Availability
  • Add to Cart