ENAH03BB30F0 Enamine

(2R)-2-amino-3-(3-methylphenyl)propanoic acid

  • MDL number MFCD01860871
  • Cas Number : 114926-39-5
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199890
Availability details for
Quantity
  • Availability
  • Add to Cart