ENAH03BB35AE Enamine

(2E)-3-cyclopropylprop-2-enoic acid

  • MDL number MFCD03002773
  • Cas Number : 60129-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-371187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-371187
Availability details for
Quantity
  • Availability
  • Add to Cart