ENAH03BB3618 Enamine

3-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid

  • MDL number MFCD00274812
  • Cas Number : 188632-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-650420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-650420
Availability details for
Quantity
  • Availability
  • Add to Cart