ENAH03BB373E Enamine

tert-butyl 5-amino-1H-indole-1-carboxylate

  • MDL number MFCD04114746
  • Cas Number : 166104-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-749073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-749073
Availability details for
Quantity
  • Availability
  • Add to Cart