ENAH03BB4190 Enamine

{8-azabicyclo[3.2.1]octan-3-yl}methanol hydrochloride

  • MDL number MFCD23701409
  • Cas Number : 1890308-93-6
  • Molecular Weight : 177.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1268214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1268214
Availability details for
Quantity
  • Availability
  • Add to Cart