ENAH03BB4F13 Enamine

8-{[(tert-butoxy)carbonyl]amino}-1,4-dioxaspiro[4.5]decane-8-carboxylic acid

  • MDL number MFCD04115504
  • Cas Number : 886362-27-2
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1272642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1272642
Availability details for
Quantity
  • Availability
  • Add to Cart