ENAH03BB4FA1 Enamine

3-(aminomethyl)cyclobutan-1-one hydrochloride

  • MDL number MFCD19707644
  • Cas Number : 1416323-22-2
  • Molecular Weight : 135.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1273190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1273190
Availability details for
Quantity
  • Availability
  • Add to Cart