ENAH03BBFBE4 Enamine

butyl(ethyl)nitrosoamine

  • MDL number MFCD00015379
  • Cas Number : 4549-44-4
  • Molecular Weight : 130.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1296532

Safety Information

signalword Danger
pcodes P201,P202,P261,P264,P270,P271,P280,P281,P301+P310,P302+P352,P304+P340,P305+P351+P338,P308+P313,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1296532
pictograms GHS06,GHS08
hstatements H301,H315,H319,H335,H350
Availability details for
Quantity
  • Availability
  • Add to Cart