ENAH03BC00E6 Enamine

(2R)-2-amino-3-(3-fluorophenyl)propanoic acid

  • MDL number MFCD00066449
  • Cas Number : 110117-84-5
  • Molecular Weight : 183.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1297344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1297344
Availability details for
Quantity
  • Availability
  • Add to Cart