ENAH03BC4A52 Enamine

(2E)-3-(pyridin-2-yl)prop-2-enoic acid

  • MDL number MFCD00179422
  • Cas Number : 54495-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1455843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1455843
Availability details for
Quantity
  • Availability
  • Add to Cart