ENAH03BC4FA3 Enamine

3-[6-(trifluoromethyl)pyridin-3-yl]propanoic acid

  • MDL number MFCD06801326
  • Cas Number : 539855-70-4
  • Molecular Weight : 219.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1457375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1457375
Availability details for
Quantity
  • Availability
  • Add to Cart