ENAH03BC509E Enamine

4-formylbenzene-1-sulfonic acid

  • MDL number MFCD22570289
  • Cas Number : 5363-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-123
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1456888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1456888
Availability details for
Quantity
  • Availability
  • Add to Cart