ENAH03BC69D4 Enamine

5,6,7,8-tetrahydroquinazolin-2-amine

  • MDL number MFCD08060043
  • Cas Number : 2305-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1462197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1462197
Availability details for
Quantity
  • Availability
  • Add to Cart