ENAH0424ABF0 Enamine

4-methylquinolin-3-amine

  • MDL number MFCD02666056
  • Cas Number : 50878-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118412
Availability details for
Quantity
  • Availability
  • Add to Cart