ENAH0424AD54 Enamine

2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)propanoic acid

  • MDL number MFCD00457538
  • Cas Number : 39137-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43125
Availability details for
Quantity
  • Availability
  • Add to Cart