ENAH0424AD7F Enamine

1-cyclopropyl-1H-pyrazole

  • MDL number MFCD22548191
  • Cas Number : 1151814-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101335
Availability details for
Quantity
  • Availability
  • Add to Cart