ENAH0424ADAB Enamine

(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanedioic acid

  • MDL number MFCD00237657
  • Cas Number : 121343-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81249
Availability details for
Quantity
  • Availability
  • Add to Cart