ENAH0424AE00 Enamine

4-oxocyclohexane-1-sulfonamide

  • MDL number MFCD30150218
  • Cas Number : 1934434-18-0
  • Molecular Weight : 177.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98090

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98090
Availability details for
Quantity
  • Availability
  • Add to Cart