ENAH0424AEEE Enamine

methyl 5-bromo-4-methyl-1H-indole-2-carboxylate

  • MDL number MFCD29035115
  • Cas Number : 1857296-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-219952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219952
Availability details for
Quantity
  • Availability
  • Add to Cart