ENAH0424AF13 Enamine

1-methyl-5-(tributylstannyl)-1H-1,2,3-triazole

  • MDL number MFCD13190023
  • Cas Number : 170681-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-244483

Safety Information

signalword Danger
pcodes P260,P264,P270,P273,P280,P301+P310,P302+P352,P305+P351+P338,P312,P314,P330,P332+P313,P337+P313,P362,P363,P391,P405,P501
hcodes N,T,Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-244483
pictograms GHS06,GHS08,GHS09
hstatements H301,H312,H315,H319,H372,H410
Availability details for
Quantity
  • Availability
  • Add to Cart