ENAH0424B29B Enamine

1-[(tert-butoxy)carbonyl]-3-ethylazetidine-3-carboxylic acid

  • MDL number MFCD12406582
  • Cas Number : 610791-06-5
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-640529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-640529
Availability details for
Quantity
  • Availability
  • Add to Cart