ENAH0424B48E Enamine

lithium(1+) ion 5-fluoropyridine-2-sulfinate

  • MDL number MFCD31615727
  • Cas Number : 2172079-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-754034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-754034
Availability details for
Quantity
  • Availability
  • Add to Cart