ENAH0424B4EE Enamine

5-tert-butylpyrimidin-2-amine

  • MDL number MFCD18260345
  • Cas Number : 94615-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-746910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-746910
Availability details for
Quantity
  • Availability
  • Add to Cart