ENAH0424B502 Enamine

3-cyclobutyl-1-methyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD31616311
  • Cas Number : 2169086-17-1
  • Molecular Weight : 234.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1166996

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1166996
Availability details for
Quantity
  • Availability
  • Add to Cart