ENAH0424B530 Enamine

trichloroethanecarbonyl isothiocyanate

  • MDL number MFCD31629063
  • Cas Number : 15146-49-3
  • Molecular Weight : 204.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1179936

Safety Information

signalword Danger
pcodes P210,P261,P264,P270,P280,P285,P301+P310,P304+P341,P305+P351+P338,P310,P330,P342+P311,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1179936
pictograms GHS05,GHS06,GHS08
hstatements H227,H301,H318,H334
Availability details for
Quantity
  • Availability
  • Add to Cart