ENAH0424B553 Enamine

(2S,4R)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-4-(2-methoxy-2-oxoethyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD31794391
  • Cas Number : 2171265-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-814159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-814159
Availability details for
Quantity
  • Availability
  • Add to Cart