ENAH0424B574 Enamine

6-bromo-3-fluoro-2-(trifluoromethyl)pyridine

  • MDL number MFCD16609979
  • Cas Number : 1227578-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-816395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-816395
Availability details for
Quantity
  • Availability
  • Add to Cart