ENAH0424B7B7 Enamine

tert-butyl 2,5-dimethyl-1-(prop-2-yn-1-yl)-1H-pyrrole-3-carboxylate

  • MDL number MFCD31629234
  • Cas Number : 2172256-43-6
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1270204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1270204
Availability details for
Quantity
  • Availability
  • Add to Cart