ENAH0424BA7E Enamine

6-bromo-5-chloro-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD28402357
  • Cas Number : 1154740-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1442289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1442289
Availability details for
Quantity
  • Availability
  • Add to Cart