ENAH0424BB06 Enamine

4-chloropyrazolo[1,5-a]pyridine-2-carboxylic acid

  • MDL number MFCD30541566
  • Cas Number : 2044704-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1478717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1478717
Availability details for
Quantity
  • Availability
  • Add to Cart