ENAH0424BB1C Enamine

1,2-oxazole-5-carbonitrile

  • MDL number MFCD18909311
  • Cas Number : 68776-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1478765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1478765
Availability details for
Quantity
  • Availability
  • Add to Cart