ENAH0424CA0B Enamine

3-nitrophenyl chloroformate

  • MDL number MFCD24141123
  • Cas Number : 14235-05-3
  • Molecular Weight : 201.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1612319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1612319
Availability details for
Quantity
  • Availability
  • Add to Cart